Assay Details
Target Gene Details
Entrez Gene ID: | 71764 |
Gene Name: | C2 calcium-dependent domain containing 2-like |
Gene Aliases: |
1300006O23Rik, Tmem24 |
Location: |
Chr.9:44309237-44320324 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd2l | NM_027909.2 | NP_082185.2 | ||
| AK004920.1 | BAB23671.1 | |||
| AK039220.1 | BAC30281.1 | |||
| AK122237.1 | 7 | 3443 | BAC65519.1 | |
| AK153804.1 | BAE32190.1 | |||
| BC035271.1 | 9 | 1874 | AAH35271.1 | |
| BC049875.1 | AAH49875.1 | |||
| BC056612.1 | AAH56612.1 | |||
| BC060156.1 | AAH60156.1 |

Genomic Map