Assay Details
Target Gene Details
Entrez Gene ID: | 13077 |
Gene Name: | cytochrome P450, family 1, subfamily a, polypeptide 2 |
Gene Aliases: |
CP12, Cyp1a1, P450-3 |
Location: |
Chr.9:57676937-57683655 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp1a2 | NM_009993.3 | NP_034123.1 | ||
| BC018298.1 | AAH18298.1 | |||
| BC054827.1 | AAH54827.1 | |||
| K02589.1 | AAA37509.1 | |||
| X00479.1 | CAA25156.1 |

Genomic Map