Assay Details
Target Gene Details
Entrez Gene ID: | 12903 |
Gene Name: | cellular retinoic acid binding protein I |
Gene Aliases: |
AI326249, CRABP-I, Crabp-1, CrabpI, Rbp-5 |
Location: |
Chr.9:54764748-54773110 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crabp1 | NM_001284507.1 | 1 | 104 | NP_001271436.1 |
| NM_013496.3 | 1 | 104 | NP_038524.1 | |
| AK045283.1 | BAC32295.1 | |||
| AK144397.1 | 1 | 56 | BAE25869.1 | |
| BC065787.1 | AAH65787.1 | |||
| X15481.1 | CAA33509.1 | |||
| X15789.1 | CAA33790.1 |

Genomic Map