Assay Details
Target Gene Details
Entrez Gene ID: | 235599 |
Gene Name: | RIKEN cDNA 6430571L13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:107340640-107349683 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6430571L13Rik | NM_175486.3 | NP_780695.1 | ||
| AK032358.1 | BAC27833.1 | |||
| AK032501.1 | BAC27899.1 | |||
| AK043338.1 | BAC31524.1 | |||
| AK085251.1 | BAC39401.1 | |||
| BC115775.1 | ||||
| BC116708.1 |
More Information
Set Membership: |
|

Genomic Map