Assay Details
Target Gene Details
Entrez Gene ID: | 78934 |
Gene Name: | RIKEN cDNA 4930581F22 gene |
Gene Aliases: |
- |
Location: |
Chr.9:35116728-35130922 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930581F22Rik | NR_029475.1 | |||
| AK019824.1 | BAB31868.1 | |||
| AW986112.1 | ||||
| CX731663.1 |
Target Gene Details
Entrez Gene ID: | 69305 |
Gene Name: | decapping enzyme, scavenger |
Gene Aliases: |
1700001E16Rik, AA408441 |
Location: |
Chr.9:35124414-35175987 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcps | NM_027030.2 | NP_081306.1 | ||
| AK005584.1 | BAB24138.1 | |||
| AK078253.1 | BAC37194.1 | |||
| AK169641.1 | ||||
| AK171018.1 | BAE42188.1 | |||
| AY040775.1 | AAK91764.1 | |||
| BC016273.1 | AAH16273.1 |
More Information
Set Membership: |
|

Genomic Map