Assay Details
Target Gene Details
Entrez Gene ID: | 75600 |
Gene Name: | calmodulin-like 4 |
Gene Aliases: |
2010002G05Rik |
Location: |
Chr.9:62857481-62875921 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calml4 | NM_001102468.1 | NP_001095938.1 | ||
| NM_138304.2 | NP_612177.1 | |||
| AK008039.1 | BAC25195.1 | |||
| AK146656.1 | BAE27336.1 | |||
| AY061807.1 | AAL38053.1 | |||
| BC013541.1 | ||||
| BC023475.1 | AAH23475.1 | |||
| BY097052.1 |
More Information
Set Membership: |
|

Genomic Map