Assay Details
Target Gene Details
Entrez Gene ID: | 30959 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 25 |
Gene Aliases: |
AW047046, GRTH |
Location: |
Chr.9:35541848-35558470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx25 | NM_013932.4 | NP_038960.2 | ||
| AF142630.3 | AAF21361.2 | |||
| AK050693.1 | BAC34384.1 | |||
| AK078340.1 | BAC37227.1 | |||
| BC024852.1 | AAH24852.1 | |||
| BC061130.1 |

Genomic Map