Assay Details
Target Gene Details
Entrez Gene ID: | 13478 |
Gene Name: | dolichyl-phosphate (UDP-N-acetylglucosamine) acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase) |
Gene Aliases: |
AU021132, Dpagt2, GPT, Gnpta |
Location: |
Chr.9:44326017-44333606 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpagt1 | NM_007875.2 | 2 | 419 | NP_031901.2 |
| AK008246.1 | BAB25555.1 | |||
| AK008317.1 | BAB25597.1 | |||
| AK164129.1 | 2 | 354 | ||
| BC003943.1 | 2 | 76 | AAH03943.1 | |
| BC010474.1 | 2 | 419 | AAH10474.1 | |
| X65603.1 | 2 | 284 | CAA46553.1 |
More Information
Set Membership: |
|

Genomic Map