Assay Details
Target Gene Details
Entrez Gene ID: | 66917 |
Gene Name: | cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1 |
Gene Aliases: |
1110001O09Rik, AA409036, Chp-1, morgana |
Location: |
Chr.9:18292233-18314000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chordc1 | NM_025844.2 | NP_080120.1 | ||
| AK003259.1 | BAB22675.1 | |||
| BC018374.1 | AAH18374.1 |
More Information
Set Membership: |
|

Genomic Map