Assay Details
Target Gene Details
Entrez Gene ID: | 270156 |
Gene Name: | expressed sequence AU019823 |
Gene Aliases: |
Gm639 |
Location: |
Chr.9:50605240-50617589 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AU019823 | NM_001134902.1 | NP_001128374.1 | ||
| NM_212449.2 | NP_997614.2 | |||
| AK133777.1 | ||||
| BC067205.1 | AAH67205.1 | |||
| BY740784.1 | ||||
| CJ142322.1 |

Genomic Map