Assay Details
Target Gene Details
Entrez Gene ID: | 12700 |
Gene Name: | cytokine inducible SH2-containing protein |
Gene Aliases: |
AI385595, CIS1, Cis, F17, F23 |
Location: |
Chr.9:107296685-107301962 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cish | NM_009895.3 | NP_034025.1 | ||
| AK077338.1 | BAC36758.1 | |||
| AK170116.1 | BAE41574.1 | |||
| AK170410.1 | BAE41776.1 | |||
| AK170554.1 | BAE41877.1 | |||
| AK170877.1 | BAE42087.1 | |||
| AK171431.1 | BAE42448.1 | |||
| BC003783.1 | AAH03783.1 | |||
| BC022178.1 | AAH22178.1 | |||
| D31943.1 | BAA06713.1 |
More Information
Set Membership: |
|

Genomic Map