Assay Details
Target Gene Details
Entrez Gene ID: | 235435 |
Gene Name: | lactase-like |
Gene Aliases: |
E130104I05Rik, KLPH |
Location: |
Chr.9:64117123-64138118 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lctl | NM_145835.2 | NP_665834.1 | ||
| AF309072.1 | AAM77699.1 |
More Information
Set Membership: |
|

Genomic Map