Assay Details
Target Gene Details
Entrez Gene ID: | 66163 |
Gene Name: | mitochondrial ribosomal protein L4 |
Gene Aliases: |
1110017G11Rik |
Location: |
Chr.9:21002605-21008839 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl4 | NM_023167.2 | 7 | 814 | NP_075656.2 |
| AB041599.1 | 6 | 566 | BAA95082.1 | |
| AB049634.1 | 7 | 813 | BAB40839.1 | |
| AK002464.1 | 7 | 650 | BAB22118.1 | |
| AK003746.2 | 7 | 771 | ||
| BC037064.1 | 7 | 647 | AAH37064.1 | |
| BC039983.1 | 7 | 782 | AAH39983.2 | |
| BC061095.1 | 7 | 630 | AAH61095.1 |
More Information
Set Membership: |
|

Genomic Map