Assay Details
Target Gene Details
Entrez Gene ID: | 54369 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
nm23-M6 |
Location: |
Chr.9:109832711-109842974 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme6 | NM_018757.1 | NP_061227.1 | ||
| AF051942.1 | AAC78464.1 | |||
| AK004201.1 | ||||
| BC002007.1 | AAH02007.1 |
More Information
Set Membership: |
|

Genomic Map