Assay Details
Target Gene Details
Entrez Gene ID: | 244911 |
Gene Name: | C2 calcium-dependent domain containing 4A |
Gene Aliases: |
EG244911, Fam148a |
Location: |
Chr.9:67830532-67832330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd4a | NM_001163143.1 | NP_001156615.1 |

Genomic Map