Assay Details
Target Gene Details
Entrez Gene ID: | 15288 |
Gene Name: | hydroxymethylbilane synthase |
Gene Aliases: |
PBGD, T25658, Ups, Uros1 |
Location: |
Chr.9:44336348-44344228 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmbs | NM_001110251.1 | 1 | 43 | NP_001103721.1 |
| NM_013551.2 | NP_038579.2 | |||
| AK028234.1 | ||||
| AK143558.1 | BAE25434.1 | |||
| AK151020.1 | BAE30038.1 | |||
| AK153884.1 | ||||
| AK158160.1 | ||||
| AK166734.1 | BAE38979.1 | |||
| AK167702.1 | BAE39746.1 | |||
| BB844509.1 | 1 | 44 | ||
| BC003861.1 | AAH03861.1 | |||
| BY786671.1 |
More Information
Set Membership: |
|

Genomic Map