Assay Details
Target Gene Details
Entrez Gene ID: | 84004 |
Gene Name: | melanoma cell adhesion molecule |
Gene Aliases: |
1-gicerin, AV025631, CD146, CD149, Muc18, s-endo, s-gicerin |
Location: |
Chr.9:44134584-44142727 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcam | NM_023061.2 | NP_075548.2 | ||
| AB035508.1 | BAB16050.1 | |||
| AB035509.1 | BAB16051.1 | |||
| BC026985.1 | AAH26985.1 | |||
| CJ045252.1 | ||||
| CN699979.1 |

Genomic Map