Assay Details
Target Gene Details
Entrez Gene ID: | 16154 |
Gene Name: | interleukin 10 receptor, alpha |
Gene Aliases: |
AW553859, CDw210, CDw210a, Il10r, mIL-10R |
Location: |
Chr.9:45253837-45269251 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il10ra | NM_008348.2 | 4 | 536 | NP_032374.1 |
| AK137663.1 | 4 | 537 | BAE23452.1 | |
| AK149680.1 | 4 | 531 | BAE29022.1 | |
| AK149899.1 | 3 | 419 | BAE29153.1 | |
| AK150022.1 | 4 | 533 | BAE29247.1 | |
| AK150524.1 | 4 | 540 | ||
| AK153120.1 | 4 | 540 | BAE31735.1 | |
| AK159566.1 | 4 | 533 | BAE35189.1 | |
| L12120.1 | 4 | 541 | AAA16156.1 |
More Information
Set Membership: |
|

Genomic Map