Assay Details
Target Gene Details
Entrez Gene ID: | 28015 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide M |
Gene Aliases: |
AA407243, AA408799, AI303746, AW553238, D9Wsu138e, Grinl1a, mKIAA4185 |
Location: |
Chr.9:71478437-71485983 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2m | NM_178602.3 | NP_848717.1 | ||
| AK014316.1 | BAB29267.1 | |||
| AK017773.1 | ||||
| AK051868.1 | BAC34793.1 | |||
| AK052037.1 | BAC34836.1 | |||
| AK078750.1 | ||||
| AK079119.1 | BAC37548.1 | |||
| AK136806.1 | ||||
| AK151221.1 | ||||
| AK159924.1 | BAE35486.1 | |||
| AK166738.1 | BAE38981.1 | |||
| BC018502.1 | AAH18502.1 | |||
| BC062199.1 | AAH62199.1 | |||
| BQ443340.1 |

Genomic Map