Assay Details
Target Gene Details
Entrez Gene ID: | 14860 |
Gene Name: | glutathione S-transferase, alpha 4 |
Gene Aliases: |
GST5.7, mGsta4 |
Location: |
Chr.9:78191966-78209349 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsta4 | NM_010357.3 | NP_034487.2 | ||
| BU953110.1 |
More Information
Set Membership: |
|

Genomic Map