Assay Details
Target Gene Details
Entrez Gene ID: | 14860 |
Gene Name: | glutathione S-transferase, alpha 4 |
Gene Aliases: |
GST5.7, mGsta4 |
Location: |
Chr.9:78191966-78209349 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsta4 | NM_010357.3 | NP_034487.2 | ||
| AI043064.1 | ||||
| AK008189.1 | BAB25520.1 | |||
| AK008193.1 | BAB25524.1 | |||
| AK008400.1 | BAB25649.1 | |||
| AK008490.1 | BAB25696.1 | |||
| AK009668.1 | BAB26429.1 | |||
| AK010098.1 | BAB26701.1 | |||
| AK011177.1 | BAB27449.1 | |||
| AK011841.1 | BAB27873.1 | |||
| AK019271.1 | BAB31640.1 | |||
| BC012639.1 | AAH12639.1 | |||
| BU953110.1 | ||||
| L06047.1 | AAA37754.1 |
More Information
Set Membership: |
|

Genomic Map