Assay Details
Target Gene Details
Entrez Gene ID: | 14012 |
Gene Name: | myelin protein zero-like 2 |
Gene Aliases: |
Eva, Eva1 |
Location: |
Chr.9:45040291-45054043 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpzl2 | NM_007962.4 | NP_031988.2 | ||
| AF030454.1 | AAC40128.1 | |||
| AK032788.1 | BAC28022.1 | |||
| AK166341.1 | BAE38716.1 | |||
| AK167263.1 | BAE39377.1 | |||
| AK170816.1 | BAE42048.1 | |||
| BC015076.1 | AAH15076.1 |
More Information
Set Membership: |
|

Genomic Map