Assay Details
Target Gene Details
Entrez Gene ID: | 319742 |
Gene Name: | myelin protein zero-like 3 |
Gene Aliases: |
5430427F17Rik, A530065I17Rik, rc |
Location: |
Chr.9:45055181-45075042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpzl3 | NM_001093749.2 | NP_001087218.1 | ||
| EF102773.1 |
More Information
Set Membership: |
|

Genomic Map