Assay Details
Target Gene Details
Entrez Gene ID: | 22041 |
Gene Name: | transferrin |
Gene Aliases: |
AI266983, Cd176, HP, Tf, Tfn, hpx |
Location: |
Chr.9:103208876-103230286 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trf | NM_133977.2 | 3 | 346 | NP_598738.1 |
| AF440692.1 | 2 | 276 | AAL34533.1 | |
| AK085754.1 | 3 | 331 | BAC39532.1 | |
| AK146549.1 | 3 | 331 | BAE27253.1 | |
| AK149559.1 | 3 | 331 | BAE28960.1 | |
| AK149595.1 | 3 | 331 | BAE28981.1 | |
| AK150782.1 | 3 | 379 | BAE29847.1 | |
| AK168368.1 | 3 | 331 | ||
| AK168405.1 | 3 | 331 | ||
| AK168419.1 | 3 | 331 | BAE40332.1 | |
| AK168938.1 | 3 | 331 | BAE40747.1 | |
| AK169187.1 | 3 | 331 | ||
| BC008559.1 | 1 | 68 | AAH08559.1 | |
| BC012313.1 | 3 | 346 | AAH12313.1 | |
| BC022986.1 | 3 | 324 | AAH22986.1 | |
| BC092046.1 | 3 | 307 | AAH92046.1 | |
| M23013.1 | 1 | 81 | AAA40488.1 |
More Information
Set Membership: |
|

Genomic Map