Assay Details
Target Gene Details
Entrez Gene ID: | 22041 |
Gene Name: | transferrin |
Gene Aliases: |
AI266983, Cd176, HP, Tf, Tfn, hpx |
Location: |
Chr.9:103208876-103230286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trf | NM_133977.2 | NP_598738.1 | ||
| AF440692.1 | AAL34533.1 | |||
| AK085754.1 | BAC39532.1 | |||
| AK146549.1 | BAE27253.1 | |||
| AK149559.1 | BAE28960.1 | |||
| AK149595.1 | BAE28981.1 | |||
| AK150782.1 | BAE29847.1 | |||
| AK168368.1 | ||||
| AK168405.1 | ||||
| AK168419.1 | BAE40332.1 | |||
| AK168938.1 | BAE40747.1 | |||
| AK169187.1 | ||||
| BC008559.1 | AAH08559.1 | |||
| BC012313.1 | AAH12313.1 | |||
| BC022986.1 | AAH22986.1 | |||
| BC092046.1 | AAH92046.1 | |||
| J03299.1 | AAA39438.1 | |||
| M23014.1 | AAA40489.1 | |||
| S67217.1 | AAB28966.2 |
More Information
Set Membership: |
|

Genomic Map