Assay Details
Target Gene Details
Entrez Gene ID: | 22273 |
Gene Name: | ubiquinol-cytochrome c reductase core protein 1 |
Gene Aliases: |
1110032G10Rik |
Location: |
Chr.9:108936648-108949641 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrc1 | NM_025407.2 | 4 | 434 | NP_079683.2 |
| AK002292.1 | 4 | 375 | ||
| AK004031.1 | 4 | 380 | ||
| AK010553.1 | 4 | 374 | BAB27022.1 | |
| AK013128.1 | 4 | 374 | BAB28666.1 | |
| AK151764.1 | 4 | 376 | BAE30670.1 | |
| AK160337.1 | 4 | 381 | BAE35744.1 | |
| AK167911.1 | 4 | 374 | BAE39918.1 | |
| AK168218.1 | 4 | 376 | BAE40175.1 | |
| AK177346.1 | 3 | 227 | ||
| AK177461.1 | 3 | 227 | ||
| AK181814.1 | 3 | 227 | ||
| AK183410.1 | 3 | 227 | ||
| AK184915.1 | 3 | 227 | ||
| AK185377.1 | 3 | 227 | ||
| BB649589.1 | 4 | 434 | ||
| BC030064.1 | 4 | 371 | AAH30064.1 | |
| BY260300.1 |
More Information
Set Membership: |
|

Genomic Map