Assay Details
Target Gene Details
Entrez Gene ID: | 258601 |
Gene Name: | olfactory receptor 984 |
Gene Aliases: |
MOR239-6 |
Location: |
Chr.9:40100544-40156029 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr984 |
Target Gene Details
Entrez Gene ID: | 75667 |
Gene Name: | transmembrane protein 225 |
Gene Aliases: |
1700030E15Rik, Tnen225 |
Location: |
Chr.9:40148100-40150901 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem225 | NM_029379.1 | 2 | 456 | NP_083655.1 |
| AK006538.1 | 2 | 457 | BAB24641.1 | |
| BC116869.1 | 2 | 372 | ||
| BC116871.1 | 2 | 372 |

Genomic Map