Assay Details
Target Gene Details
Entrez Gene ID: | 70348 |
Gene Name: | ubiquitin-conjugating enzyme E2C binding protein |
Gene Aliases: |
2610018I03Rik, Ube3d |
Location: |
Chr.9:86307234-86465181 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2cbp | NM_027394.2 | NP_081670.1 | ||
| AK011446.1 | BAB27624.1 | |||
| AK049229.1 | BAC33623.1 | |||
| BC115686.1 | ||||
| BC115687.1 |
More Information
Set Membership: |
|

Genomic Map