Assay Details
Target Gene Details
Entrez Gene ID: | 22619 |
Gene Name: | sialic acid acetylesterase |
Gene Aliases: |
LSE, Ysg2 |
Location: |
Chr.9:37555637-37649054 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Siae | AF156856.1 | AAD55976.1 |
Target Gene Details
Entrez Gene ID: | 20686 |
Gene Name: | sperm autoantigenic protein 17 |
Gene Aliases: |
Sp17 |
Location: |
Chr.9:37603294-37613833 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spa17 | NM_011449.2 | NP_035579.1 | ||
| AK005571.1 | BAB24131.1 | |||
| BC059727.1 | 5 | 505 | AAH59727.1 | |
| BU936068.1 | ||||
| CK137444.1 | ||||
| Z46299.1 | CAA86455.1 |
More Information
Set Membership: |
|

Genomic Map