Assay Details
Target Gene Details
Entrez Gene ID: | 30057 |
Gene Name: | translocase of inner mitochondrial membrane 8B |
Gene Aliases: |
Tim8b |
Location: |
Chr.9:50603901-50605320 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Timm8b | NM_013897.2 | NP_038925.1 | ||
| AA204614.1 | ||||
| AK003382.1 | BAB22753.1 | |||
| AK012925.1 | BAB28552.1 | |||
| AK018718.1 | BAB31364.1 | |||
| AK019422.1 | BAB31713.1 | |||
| AK169258.1 | BAE41021.1 | |||
| BC029239.1 | AAH29239.1 | |||
| BY100901.1 |

Genomic Map