Assay Details
Target Gene Details
Entrez Gene ID: | 72229 |
Gene Name: | RIKEN cDNA 1700003G13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:45318266-45322079 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700003G13Rik | NR_040720.1 | |||
| AK005635.1 |
Target Gene Details
Entrez Gene ID: | 214531 |
Gene Name: | transmembrane protease, serine 13 |
Gene Aliases: |
BC010843, Tpmrss13 |
Location: |
Chr.9:45319071-45347581 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss13 | NM_001013373.2 | NP_001013391.2 | ||
| BC085323.1 | AAH85323.1 |
More Information
Set Membership: |
|

Genomic Map