Assay Details
Target Gene Details
Entrez Gene ID: | 78709 |
Gene Name: | serine peptidase inhibitor, Kazal type 8 |
Gene Aliases: |
C630041L24Rik |
Location: |
Chr.9:109812203-109826628 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink8 | NM_183136.2 | 1 | 97 | NP_898959.2 |
| AK021276.1 | 1 | 80 | BAC25651.1 | |
| BC048588.1 | 1 | 97 | AAH48588.1 | |
| CN843460.1 | 1 | 43 | ||
| DQ437329.1 | 1 | 79 |
More Information
Set Membership: |
|

Genomic Map