Assay Details
Target Gene Details
Entrez Gene ID: | 66131 |
Gene Name: | timeless interacting protein |
Gene Aliases: |
1110005A05Rik, 1110018P21Rik |
Location: |
Chr.9:64281489-64304794 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tipin | NM_025372.3 | 5 | 386 | NP_079648.1 |
| AK003451.1 | 5 | 380 | BAB22798.1 | |
| AK003802.1 | 5 | 387 | BAB23004.1 | |
| AK011357.1 | 5 | 381 | BAB27565.1 | |
| BC016211.1 | 4 | 363 | AAH16211.1 |
More Information
Set Membership: |
|

Genomic Map