Assay Details
Target Gene Details
Entrez Gene ID: | 117149 |
Gene Name: | toll-interleukin 1 receptor (TIR) domain-containing adaptor protein |
Gene Aliases: |
AA407980, C130027E04Rik, Mal, Tlr4ap, Wyatt |
Location: |
Chr.9:35184391-35200291 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tirap | NM_001177845.1 | NP_001171316.1 | ||
| NM_001177846.1 | NP_001171317.1 | |||
| NM_001177847.1 | NP_001171318.1 | |||
| NM_054096.2 | NP_473437.2 | |||
| AF378130.1 | AAL05628.1 | |||
| AK036431.1 | BAC29426.1 | |||
| AK080135.1 | BAC37831.1 | |||
| AK151117.1 | BAE30126.1 | |||
| AK168494.1 | BAE40381.1 | |||
| BY740695.1 | ||||
| BY743490.1 |

Genomic Map