Assay Details
Target Gene Details
Entrez Gene ID: | 234967 |
Gene Name: | solute carrier family 36 (proton/amino acid symporter), member 4 |
Gene Aliases: |
6330573I15Rik, PAT4 |
Location: |
Chr.9:15709769-15742388 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc36a4 | NM_172289.4 | 7 | 632 | NP_758493.2 |
| AF453746.1 | 8 | 631 | AAN76274.1 | |
| AK032074.1 | 7 | 633 | BAC27683.1 | |
| AK052277.1 | 7 | 633 | ||
| AK150167.1 | 7 | 664 | ||
| BC115964.1 | 7 | 633 |
More Information
Set Membership: |
|

Genomic Map