Assay Details
Target Gene Details
Entrez Gene ID: | 170761 |
Gene Name: | PDZ domain containing 3 |
Gene Aliases: |
NaPi-Cap2, Pdzk2 |
Location: |
Chr.9:44247312-44251464 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdzd3 | NM_133226.2 | NP_573489.2 | ||
| AF334612.1 | AAK20865.1 | |||
| AK052748.1 | BAC35130.1 |

Genomic Map