Assay Details
Target Gene Details
Entrez Gene ID: | 71989 |
Gene Name: | RNA pseudouridylate synthase domain containing 4 |
Gene Aliases: |
2410001E19Rik |
Location: |
Chr.9:35267881-35275957 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpusd4 | NM_028040.2 | 5 | 735 | NP_082316.1 |
| AK010322.1 | 5 | 752 | BAB26852.1 | |
| AK156574.1 | 5 | 752 | BAE33761.1 | |
| BC061256.1 | 5 | 736 | AAH61256.1 |
More Information
Set Membership: |
|

Genomic Map