Assay Details
Target Gene Details
Entrez Gene ID: | 75258 |
Gene Name: | RIKEN cDNA 4930563M21 gene |
Gene Aliases: |
Rfpl3s |
Location: |
Chr.9:55962587-56026518 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930563M21Rik | NM_183111.2 | NP_898934.2 | ||
| AK016210.1 | BAC25476.1 |

Genomic Map