Assay Details
Target Gene Details
Entrez Gene ID: | 69519 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
1700030C20Rik, AI848608, Rwdd2 |
Location: |
Chr.9:86571988-86574899 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rwdd2a | NM_001145968.1 | 2 | 222 | NP_001139440.1 |
| NM_001145969.1 | 2 | 256 | NP_001139441.1 | |
| NM_027100.2 | 2 | 351 | NP_081376.2 | |
| AK006533.1 | 2 | 350 | BAB24638.1 | |
| BC115691.1 | 2 | 351 | ||
| BC115692.1 | 2 | 222 | ||
| CJ149958.1 | 2 | 256 | ||
| CN836837.1 | 2 | 259 |
More Information
Set Membership: |
|

Genomic Map