Assay Details
Target Gene Details
Entrez Gene ID: | 77798 |
Gene Name: | RIKEN cDNA A930009A15 gene |
Gene Aliases: |
- |
Location: |
Chr.10:115569809-115582454 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A930009A15Rik | NM_029982.1 | NP_084258.1 | ||
| AK020834.1 |
Target Gene Details
Entrez Gene ID: | 14160 |
Gene Name: | leucine rich repeat containing G protein coupled receptor 5 |
Gene Aliases: |
FEX, Gpr49 |
Location: |
Chr.10:115450314-115587780 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgr5 | NM_010195.2 | NP_034325.2 | ||
| AF110818.1 | AAD14684.1 | |||
| AK047873.1 | BAC33180.1 | |||
| AK132387.1 | BAE21138.1 | |||
| AK134981.1 |
More Information
Set Membership: |
|

Genomic Map