Assay Details
Target Gene Details
Entrez Gene ID: | 212862 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:88472813-88503970 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chpt1 | NM_001146690.1 | NP_001140162.1 | ||
| NR_027477.1 | ||||
| AK007964.1 | ||||
| AW823530.1 | ||||
| BE137041.1 |
Target Gene Details
Entrez Gene ID: | 20962 |
Gene Name: | synaptonemal complex protein 3 |
Gene Aliases: |
Cor1, Scp3 |
Location: |
Chr.10:88459587-88473236 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sycp3 | NM_011517.2 | 9 | 856 | NP_035647.2 |
| AK131949.1 | 9 | 780 | BAE20894.1 | |
| BC132079.1 | 9 | 857 | ||
| BC138509.1 | 9 | 857 | ||
| BY386732.1 | 2 | 155 | ||
| Y08485.1 | 9 | 865 | CAA69719.1 |
More Information
Set Membership: |
|

Genomic Map