Assay Details
Target Gene Details
Entrez Gene ID: | 58250 |
Gene Name: | carbohydrate sulfotransferase 11 |
Gene Aliases: |
1110020P09Rik, C4ST, C4ST-1, C4ST1, C4s |
Location: |
Chr.10:82985380-83195907 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chst11 | NM_021439.2 | NP_067414.2 | ||
| AB030378.1 | BAA92942.1 | |||
| AJ289133.1 | CAB92141.1 | |||
| BC137629.1 | ||||
| BC144793.1 |
More Information
Set Membership: |
|

Genomic Map