Assay Details
Target Gene Details
Entrez Gene ID: | 52717 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
2310005G07Rik, APC16, D10Ertd641e |
Location: |
Chr.10:59987409-60003131 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc16 | NM_025514.2 | NP_079790.1 | ||
| AK009172.1 | BAB26121.1 | |||
| AK012280.1 | BAB28138.1 | |||
| AK088376.1 | BAC40315.1 |
Target Gene Details
Entrez Gene ID: | 69090 |
Gene Name: | activating signal cointegrator 1 complex subunit 1 |
Gene Aliases: |
1810015P09Rik, AI550520, ASC1p50, CGI-18 |
Location: |
Chr.10:60002493-60099990 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascc1 | NM_026937.3 | NP_081213.1 | ||
| AK053811.1 | ||||
| AK144614.1 | BAE25970.1 | |||
| AK171958.1 | BAE42747.1 | |||
| BB844213.1 | ||||
| BC030905.1 | AAH30905.1 |
More Information
Set Membership: |
|

Genomic Map