Assay Details
Target Gene Details
Entrez Gene ID: | 68778 |
Gene Name: | guanylyl cyclase domain containing 1 |
Gene Aliases: |
1110038D17Rik |
Location: |
Chr.10:75506814-75517967 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucd1 | NM_175133.1 | 3 | 342 | NP_780342.1 |
| AK088820.1 | 3 | 287 | ||
| AK157404.1 | 3 | 401 | BAE34080.1 | |
| AK170872.1 | 3 | 378 | ||
| AK180276.1 | ||||
| BC024851.1 | 3 | 296 | ||
| BC043463.1 | 3 | 342 | ||
| BC118613.1 | 3 | 400 | ||
| BC119795.1 | 3 | 400 |
More Information
Set Membership: |
|

Genomic Map