Assay Details
Target Gene Details
Entrez Gene ID: | 71712 |
Gene Name: | DNA-damage regulated autophagy modulator 1 |
Gene Aliases: |
1200002N14Rik, Dram |
Location: |
Chr.10:88322801-88364383 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dram1 | NM_027878.2 | NP_082154.2 | ||
| AK004552.1 | BAB23366.2 | |||
| AK151184.1 | ||||
| BC021433.1 | AAH21433.1 |
More Information
Set Membership: |
|

Genomic Map