Assay Details
Target Gene Details
Entrez Gene ID: | 74007 |
Gene Name: | BTB (POZ) domain containing 11 |
Gene Aliases: |
6330404E16Rik |
Location: |
Chr.10:85386776-85660292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btbd11 | NM_001017525.1 | NP_001017525.1 | ||
| NM_028709.2 | NP_082985.2 | |||
| AK018115.1 | BAB31077.1 | |||
| AK039038.1 | BAC30216.1 | |||
| AK044022.1 | BAC31741.1 | |||
| BC059036.1 | AAH59036.1 | |||
| BC072592.1 | AAH72592.1 | |||
| BC141145.1 |
More Information
Set Membership: |
|

Genomic Map