Assay Details
Target Gene Details
Entrez Gene ID: | 52717 |
Gene Name: | anaphase promoting complex subunit 16 |
Gene Aliases: |
2310005G07Rik, APC16, D10Ertd641e |
Location: |
Chr.10:59987409-60003131 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc16 | NM_025514.2 | NP_079790.1 | ||
| AK009172.1 | BAB26121.1 | |||
| AK012280.1 | BAB28138.1 | |||
| AK088376.1 | BAC40315.1 |

Genomic Map