Assay Details
Target Gene Details
Entrez Gene ID: | 67454 |
Gene Name: | IKBKB interacting protein |
Gene Aliases: |
1200009F10Rik, 1700023M03Rik, D18354, Ikip |
Location: |
Chr.10:91083039-91102613 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbip | NM_026166.2 | 2 | 325 | NP_080442.2 |
| NM_027078.2 | 2 | 325 | NP_081354.1 | |
| AK004670.1 | 2 | 283 | BAB23459.1 | |
| AK006283.1 | 2 | 293 | BAB24502.1 | |
| BC034890.1 | 2 | 287 | AAH34890.1 | |
| BC055834.1 | 2 | 276 | AAH55834.1 | |
| BY277133.1 | 2 | 357 | ||
| CF171082.1 | 2 | 307 |

Genomic Map