Assay Details
Target Gene Details
Entrez Gene ID: | 11634 |
Gene Name: | autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy) |
Gene Aliases: |
- |
Location: |
Chr.10:78030022-78043610 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |

Genomic Map