Assay Details
Target Gene Details
Entrez Gene ID: | 216152 |
Gene Name: | lipid phosphate phosphatase-related protein type 3 |
Gene Aliases: |
Prg2 |
Location: |
Chr.10:79860475-79874634 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lppr3 | NM_001170935.1 | NP_001164406.1 | ||
| NM_181681.2 | NP_859009.2 | |||
| NR_033210.1 | ||||
| AF541278.1 | AAP57767.1 | |||
| AK083932.1 | ||||
| AK144442.1 | ||||
| AK220474.1 | BAD90500.1 | |||
| BC066006.1 | AAH66006.1 | |||
| BC098099.1 | AAH98099.1 |
More Information
Set Membership: |
|

Genomic Map